2-Bromo-3-chlorophenol,98% 1 * 5 g
Catalog # AMBDA238799-5G
Supplier: AMBEED
Frequently Bought Together
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
PROLUME NANOFUEL SOLVENT OPTIMIZED FOR IN VITRO USE WITH CTZ NATIVE H-CTZ CTZ-F AND CTZ 400 A 1 * 10 mL
Supplier: MARKET SOURCE PART PROCESS
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Supplier: PanReac AppliChem
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Turpentine oil
Supplier: Roth Carl
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone 99.6% ACS
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals