tert-Butyl (4-formylpyridin-2-yl)carbamate,95% 1 * 0,25 g
Catalog # AMBDA111117-250MG
Supplier: AMBEED
tert-Butyl (4-formylpyridin-2-yl)carbamate,95% 1 * 0,25 g
Catalog # AMBDA111117-250MG
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Chloroform, LiChrosolv® for HPLC
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Isobutanol ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Triethylamine for HPLC
Supplier: Thermo Fisher Scientific
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals