9-Bromononanoic acid,98% 1 * 1 g
Catalog # AMBDA296554-1G
Anbieter: AMBEED
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Ethanol, GPR RECTAPUR®
Anbieter: VWR Chemicals
Water, GPR RECTAPUR®
Anbieter: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Anbieter: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Anbieter: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Anbieter: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Anbieter: Merck
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Anbieter: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Anbieter: VWR Chemicals