2-Amino-5-bromobenzamide,98% 1 * 1 g
Katalognr. AMBDA228146-1G
Anbieter: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, TECHNICAL, demineralised
Anbieter: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Anbieter: Merck
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Anbieter: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Anbieter: Merck
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Anbieter: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
n-Hexane, EMSURE® ACS for analysis, Supelco®
Anbieter: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals