2-(Dimethylamino)acetyl chloride hydrochloride,95% 1 * 5 g
Catalog # AMBDA689408-5G
Supplier: AMBEED
2-(Dimethylamino)acetyl chloride hydrochloride,95% 1 * 5 g
Catalog # AMBDA689408-5G
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Videojet Technologies, OPLOSMIDDEL 750 M
Supplier: MARKET SOURCE PART PROCESS
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
p-Xylene 99% for HPLC
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1-Butanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Fisher Scientific