Hexyltrimethylammonium Bromide,98% 1 * 5 g
Catalog # AMBDA656820-5G
Supplier: AMBEED
Frequently Bought Together
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
1,4-Dioxane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific