Ethyl 3-amino-4-pyridinecarboxylate,98% 1 * 5 g
Catalog # AMBDA360871-5G
Anbieter: AMBEED
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Anbieter: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Anbieter: VWR Chemicals
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Anbieter: VWR Chemicals
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Anbieter: Merck
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Anbieter: VWR Chemicals
o-Xylene for synthesis, Sigma-Aldrich®
Anbieter: Merck
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Anbieter: VWR Chemicals