5-Acetyl-2-bromopyridine,97% 1 * 1 g
Catalog # AMBDA171418-1G
Anbieter: AMBEED
Frequently Bought Together
N,N-Dimethylacetamide 99.99-100.00%, PESTINORM® for headspace gas chromatography
Anbieter: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Anbieter: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Anbieter: VWR Chemicals