Benzyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate,96% 1 * 0,1 g
Catalog # AMBDA303963-100MG
Supplier: AMBEED
Benzyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate,96% 1 * 0,1 g
Catalog # AMBDA303963-100MG
Supplier: AMBEED
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
N-Ethyldiisopropylamine 99%
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane ≥99% inhibitor-free, Reagent Grade
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99.9% (by GC) for UV and IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethylenediamine 99+%
Supplier: Thermo Fisher Scientific
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
2-Propanol 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific