3-Bromo-4-iodonitrobenzene,98% 1 * 1 g
Catalog # AMBDA357074-1G
Supplier: AMBEED
Frequently Bought Together
N,N-Dimethylformamide, Supelco®
Supplier: Merck
Hydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Petroleum spirit, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…65 °C, Technical Grade
Supplier: Thermo Fisher Scientific
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
Vaseline Oil (USP, BP, Ph. Eur.) pure, pharma grade 1 * 5 L
Supplier: PanReac AppliChem
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Heptane, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dimethyl Sulfoxide, 99.5% for synthesis 1 * 2,5 L
Supplier: PanReac AppliChem