Methyl thiophene-3-carboxylate,98% 1 * 1 g
Catalog # AMBDA142735-1G
Supplier: AMBEED
Frequently Bought Together
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Water, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium bromide 99+% for IR spectrophotometry
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific