2-(4-Chlorophenyl)acetyl chloride,98% 1 * 5 g
Catalog # AMBDA805058-5G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% for HPLC
Supplier: Thermo Fisher Scientific
Water, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Methanol 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND