Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate,95% 1 * 0,25 g
Catalog # AMBDA660525-250MG
Supplier: AMBEED
Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate,95% 1 * 0,25 g
Catalog # AMBDA660525-250MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Methanol, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N,N-Dimethylhydrazine
Supplier: Merck
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethyl acetate, EMPLURA®, Supelco®
Supplier: Merck
1-Butanol ≥99.9%
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific