1-(4-Thiazolyl)ethanone,98% 1 * 1 g
Catalog # AMBDA401670-1G
Supplier: AMBEED
Frequently Bought Together
Methanol anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
2-Methylpentane ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dimethyl sulphoxide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals