3,6,9,12,15-Pentaoxaheptadecane-1,17-diol,97% 1 * 10 g
Catalog # AMBDA195221-10G
Supplier: AMBEED
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Methanol anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Glycerine
Supplier: Roth Carl
Nitromethane 96% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane stabilized with 20 ppm of amylene for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
1-Propanol, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Sodium hexanitritocobaltate(III)
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific