γ-Cyclodextrin,98% 1 * 100 g
Catalog # AMBDA313809-100G
Supplier: AMBEED
Frequently Bought Together
SIGTECH Mek Solvent 5 L Kanister 1 * 5 L
Supplier: MARKET SOURCE PART PROCESS
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, pure, demineralised
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Methanol ≥99.9% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.8% for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Liquid for manometer with sloping tube
Supplier: VWR Chemicals
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals
Phenazone 99%
Supplier: Thermo Fisher Scientific