4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione,98% 1 * 0,1 g
Catalog # AMBDA386017-100MG
Anbieter: AMBEED
4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione,98% 1 * 0,1 g
Catalog # AMBDA386017-100MG
Anbieter: AMBEED
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Anbieter: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Anbieter: Merck
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Anbieter: PanReac AppliChem
Chloroform ≥99.5% stabilised
Anbieter: Honeywell Chemicals
Toluene 99.8+% for analysis
Anbieter: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Anbieter: Honeywell Chemicals
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Anbieter: Avantor
N,N-Dimethylacetamide 99.5+% for analysis
Anbieter: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Anbieter: PanReac AppliChem
Methyl ethyl ketone 99+% for spectroscopy
Anbieter: Thermo Fisher Scientific
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Anbieter: Honeywell Chemicals
2-Butoxyethanol ≥99%
Anbieter: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Anbieter: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Anbieter: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Anbieter: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Anbieter: Honeywell Chemicals