Ethyl 3-ethyl-1H-pyrazole-4-carboxylate,97% 1 * 1 g
Catalog # AMBDA448116-1G
Anbieter: AMBEED
Frequently Bought Together
Methanol anhydrous, 99.8 % 1 * 1 L
Anbieter: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Anbieter: MERCK PRODUCTION CHEMICALS
1-Acetyl-2-phenylhydrazine for synthesis, Sigma-Aldrich®
Anbieter: Merck
Pyridine, extra dry 99.5%, AcroSeal®
Anbieter: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Anbieter: VWR Chemicals
Diisopropyl ether stabilised for analysis
Anbieter: Thermo Fisher Scientific
Benzyl alcohol, extra dry 98+%, AcroSeal®
Anbieter: Thermo Fisher Scientific
Glycerine 99+%, extra pure
Anbieter: Thermo Fisher Scientific
1-Butanol NF
Anbieter: Spectrum Chemical
2-Propanol, extra dry over Molecular Sieve 99.5%, AcroSeal®
Anbieter: Thermo Fisher Scientific
Butanone (Methylethylketone) pure 1 * 2,5 L
Anbieter: PanReac AppliChem
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Anbieter: Avantor
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Anbieter: Avantor