(3R,6R)-3,6-Dimethyl-1,4-dioxane-2,5-dione,98% 1 * 25 g
Catalog # AMBDA178884-25G
Supplier: AMBEED
Frequently Bought Together
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
1-Pentanol ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Potassium bromide 99+% for IR spectrophotometry
Supplier: Thermo Fisher Scientific