Dibutyryl-cAMP sodium salt,98% 1 * 0,1 g
Catalog # AMBDA976872-100MG
Supplier: AMBEED
Frequently Bought Together
PROLUME NANOFUEL SOLVENT OPTIMIZED FOR IN VITRO USE WITH CTZ NATIVE H-CTZ CTZ-F AND CTZ 400 A 1 * 10 mL
Supplier: MARKET SOURCE PART PROCESS
Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade
Supplier: PanReac AppliChem
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Isoamyl alcohol mixture of isomers ≥99%
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific