(3-Cyano-5-fluorophenyl)boronic acid,97% 1 * 5 g
Catalog # AMBDA197102-5G
Supplier: AMBEED
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Dimethyl sulphoxide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific
Nitromethane 96% for HPLC
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific