(R)-Octan-2-yl 4-((4-(hexyloxy)benzoyl)oxy)benzoate,97% 1 * 1 g
Catalog # AMBDA124771-1G
Supplier: AMBEED
(R)-Octan-2-yl 4-((4-(hexyloxy)benzoyl)oxy)benzoate,97% 1 * 1 g
Catalog # AMBDA124771-1G
Supplier: AMBEED
Frequently Bought Together
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetic acid 99-100% for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals