7-Nitroquinoline,98% 1 * 5 g
Catalog # AMBDA722991-5G
Supplier: AMBEED
Frequently Bought Together
o-Xylene anhydrous, 97 % 1 * 2 L
Supplier: Merck
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
PROLUME NANOFUEL SOLVENT OPTIMIZED FOR IN VITRO USE WITH CTZ NATIVE H-CTZ CTZ-F AND CTZ 400 A 1 * 10 mL
Supplier: MARKET SOURCE PART PROCESS
Methyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Sodium hexanitritocobaltate(III)
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific