(R)-1,2,3,4-Tetrahydronaphthoic acid,98% 1 * 0,25 g
Catalog # AMBDA310787-250MG
Supplier: AMBEED
Frequently Bought Together
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers), BAKER, J.T.Baker®
Supplier: Avantor
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific