tert-Butyl methyl(3-(oxiran-2-yl)propyl)carbamate,95% 1 * 1 g
Catalog # AMBDA1034084-1G
Supplier: AMBEED
tert-Butyl methyl(3-(oxiran-2-yl)propyl)carbamate,95% 1 * 1 g
Catalog # AMBDA1034084-1G
Supplier: AMBEED
Frequently Bought Together
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Heptane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Butyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
1-Butanol 99.5% for analysis
Supplier: Thermo Fisher Scientific
1,4-Dioxane, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific