tert-Butyl (R)-4,5-diamino-5-oxopentanoate monohydrochloride,97% 1 * 25 g
Catalog # AMBDA965113-25G
Supplier: AMBEED
tert-Butyl (R)-4,5-diamino-5-oxopentanoate monohydrochloride,97% 1 * 25 g
Catalog # AMBDA965113-25G
Supplier: AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Cyclohexane, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
2-Propanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform
Supplier: Roth Carl
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals