(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride,98% 1 * 1 g
Catalog # AMBDA1002111-1G
Supplier: AMBEED
(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride,98% 1 * 1 g
Catalog # AMBDA1002111-1G
Supplier: AMBEED
Frequently Bought Together
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Ethylene glycol, anhydrous 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Methyl formate 98% for spectroscopy
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals