2-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine,95% 1 * 0,1 g
Catalog # AMBDA986283-100MG
Supplier: AMBEED
2-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine,95% 1 * 0,1 g
Catalog # AMBDA986283-100MG
Supplier: AMBEED
Frequently Bought Together
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade
Supplier: Honeywell Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific