3-Bromopropyl acetate,95% 1 * 25 g
Catalog # AMBDA267787-25G
Supplier: AMBEED
Frequently Bought Together
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
N,N-Dimethylformamide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, Biograde for DNA and peptides synthesis
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals