ContactUsLinkComponent
AC254-125-B-ML - f=125 mm, Ø1 Achromatic Doublet, SM1-Threaded Mount, ARC: 650-1050 nm 1 * 1 ST
Catalog # THORAC254-125-B-ML
Supplier:  THORLABS
AC254-125-B-ML - f=125 mm, Ø1 Achromatic Doublet, SM1-Threaded Mount, ARC: 650-1050 nm 1 * 1 ST
Catalog # THORAC254-125-B-ML
Supplier:  THORLABS
Frequently Bought Together