Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Volumetric flasks, USP, BLAUBRAND®, class A, DE-M, 1.000 ml, NS 24/29, Boro 3.3, with PP stopper, USP individual certificate 1 * 1 items
Catalog # BRND956953
Supplier:
Brand
Expand image
Volumetric flasks, USP, BLAUBRAND®, class A, DE-M, 1.000 ml, NS 24/29, Boro 3.3, with PP stopper, USP individual certificate 1 * 1 items
Catalog # BRND956953
Supplier:
Brand
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Hydrochloric acid 32%, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection