1-{BICYCLO[2.2.0]HEXAN-1-YL}METHANAMINE 1 * 1 g
Catalog # UKROCSC015680838-1
Supplier: UKRORGSYNTHESIS
1-{BICYCLO[2.2.0]HEXAN-1-YL}METHANAMINE 1 * 1 g
Catalog # UKROCSC015680838-1
Supplier: UKRORGSYNTHESIS
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Methyl ethyl ketone ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane stabilised Ph. Eur.
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals