PROPYLENE GLYCOL MONOCAPRYLATE CAPRYOL P 1 * 25 kg
Catalog # CMPP100503
Supplier: CUSTOM MADE CHEMICALS
PROPYLENE GLYCOL MONOCAPRYLATE CAPRYOL P 1 * 25 kg
Catalog # CMPP100503
Supplier: CUSTOM MADE CHEMICALS
Frequently Bought Together
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Pyridine-2,6-dicarboxylic acid, Supelco®
Supplier: Merck
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Hydrogen chloride 1.25 mol/l, Supelco®
Supplier: Merck
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Carbon disulphide 99.9%, extra pure low in benzene
Supplier: Thermo Fisher Scientific