M 2125 CS LABRAFIL 1 * 25 kg
Catalog # CMPP101145
Supplier: CUSTOM MADE CHEMICALS
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals