ContactUsLinkComponent
One tray of 96 biosensors coated with anti-human IgG for both quantitation and kinetic analysis of human IgGs and IgG-derived Fc-fusion proteins. 1 * 1 items
Catalog # PALF18-5142
Supplier:  PALL FORTEBIO
One tray of 96 biosensors coated with anti-human IgG for both quantitation and kinetic analysis of human IgGs and IgG-derived Fc-fusion proteins. 1 * 1 items
Catalog # PALF18-5142
Supplier:  PALL FORTEBIO
Frequently Bought Together