ContactUsLinkComponent
This wafer transport cart was designed to transport 6 Wafers in wafer boxes. 9 nests that hold boxes securely and at angle all reduce wafer chatter . Anti-vibration system with dual ESD locking casters. Fabrication material: stainless steel 304, wi 1 * 1 items
Catalog # PALBCW1003
Supplier:  Palbam
undefined
This wafer transport cart was designed to transport 6 Wafers in wafer boxes. 9 nests that hold boxes securely and at angle all reduce wafer chatter . Anti-vibration system with dual ESD locking casters. Fabrication material: stainless steel 304, wi 1 * 1 items
Catalog # PALBCW1003
Supplier:  Palbam
Frequently Bought Together