ContactUsLinkComponent
Chromatograpy-columns, ST-socket 29/32, valve and PTFE-valve plug, screw-thread retaining nut, fused-in sintered glass-disc P0, 30 x 400 mm, 280 ml, bore: 2.5 mm 1 * 1 items
Catalog # WITG2.134.011
Supplier:  witeg Labortechnik
Chromatograpy-columns, ST-socket 29/32, valve and PTFE-valve plug, screw-thread retaining nut, fused-in sintered glass-disc P0, 30 x 400 mm, 280 ml, bore: 2.5 mm 1 * 1 items
Catalog # WITG2.134.011
Supplier:  witeg Labortechnik
Frequently Bought Together