ContactUsLinkComponent
Value Plastics® Fitting, Polypropylene, Straight, Male Luer to Female Luer Coupler Adapter; 1000/PK
Value Plastics® Fitting, Polypropylene, Straight, Male Luer to Female Luer Coupler Adapter; 1000/PK
Catalog # MFLX45508-82BLK
undefined
Value Plastics® Fitting, Polypropylene, Straight, Male Luer to Female Luer Coupler Adapter; 1000/PK
Catalog # MFLX45508-82BLK
About this item
Full material traceability to ensure process integrity.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of options including luer, thread, and hose barb.
  • Connection Type:
    Female Luer to Male Luer
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Male Luer to Female Luer Coupler Adapter; 1000/PK
  • Seq:
    03
  • Environmentally Preferable:
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Male luer lock
  • No. of ports:
    2
  • Pk:
    1.000
  • Fitting material:
    Polypropylene
Frequently Bought Together