AC150-A25B 13-21L Circulating Bath, -25 to 150C, 115V 1 * 1 items
Catalog # HAKALT1554258
Supplier: THERMO ELECTRON LED
AC150-A25B 13-21L Circulating Bath, -25 to 150C, 115V 1 * 1 items
Catalog # HAKALT1554258
Supplier: THERMO ELECTRON LED
Frequently Bought Together
12-CHANNEL VOYAGER ELECTRONIC PIPETTE 5- 1 * 1 items
Supplier: INTEGRA
VWR®, Accessories for VWR® Pure Water Systems
Supplier: VWR Collection
VWR® Micro Star 30, Microcentrifuge, High Speed, Universal, Ventilated
Supplier: VWR Collection
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Supplier: GILSON
Accessories for Eppendorf microcentrifuge
Supplier: EPPENDORF
Food probes
Supplier: Testo
Probes for 845
Supplier: Testo
Probes for 112
Supplier: Testo
Temperature probes, Lemo® compact, BOLA
Supplier: Bohlender
FULL SIZE HYGIENIC USB MOUSE 1 * 1 items
Supplier: TEKNOMEK
ROTOR ANGLE 44X1.5/2.0ML ID V2 1 * 1 items
Supplier: OHAUS
Ultra-accurate probes
Supplier: EBRO
Spezialadapter für Rotationsverdampfer f 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Water penetration tester, acc. to Karsten
Supplier: Industrial Physics
VWR® Traceable®, Fast Response Digital Thermometer
Supplier: VWR Collection
FILTER FOR MIELE 1 * 1 items
Supplier: MIELE