ContactUsLinkComponent
Stericlin® flat tube, 70g/m² paper and transparent film, with STEAM, EO and FORM indicators, width 10cm x length 100m, DIN EN ISO, double packed 1 * 150 Roll
Catalog # VEPA3FKFS230306A
Stericlin® flat tube, 70g/m² paper and transparent film, with STEAM, EO and FORM indicators, width 10cm x length 100m, DIN EN ISO, double packed 1 * 150 Roll
Catalog # VEPA3FKFS230306A
Frequently Bought Together