Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ANTIB ONLINE COMP antibody (Cartilage Oligomeric Matrix Protein) Internal Region 1 * 100 µl
Catalog # MSPPABIN7094624
Supplier:
MARKET SOURCE PART PROCESS
Expand image
ANTIB ONLINE COMP antibody (Cartilage Oligomeric Matrix Protein) Internal Region 1 * 100 µl
Catalog # MSPPABIN7094624
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF