SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
GM1075 SU-8 PHOTORESIST 1 LITER 1 * 1 ST
MSPPGM10751000
:
MARKET SOURCE PART PROCESS
GM1075 SU-8 PHOTORESIST 1 LITER 1 * 1 ST
MSPPGM10751000
:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
pH indicator papers, MQuant®, Supelco®
: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, copper cell test, Spectroquant®, Supelco®
: Merck
Visual colour kits, pH indicator with colour card, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Reagents for DR 2800, DR 3800 DR 3900, DR 5000 and DR 6000
: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
: Hach
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Reagents for hydrazine determination using Lovibond® MD photometers
: Lovibond