FISCHER ELEKTRONIK POWDER ELECTRODE PE 01 POWDER ELECTRODE FOR VOLUME RESISTANCE (LAYER THICKNESS 1-50 MM), INSULATING BODY PTFE, IN CLUDING ALLEN KEY 1 * 1 ST
Catalog # MSPP91140080
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together