ContactUsLinkComponent
PE-Labeled Recombinant Protein L, His Tag (RPL-PP2H2) is produced via site-specific conjugation of PE to Recombinant Protein L, His  Tag under optimal conditions with a proprietary technology. Recombinant Protein L is expressed from E.coli cells. It 1 * 200 Tests
Catalog # ACRBRPL-PP2H2-200
Supplier:  ACRO Biosystems
PE-Labeled Recombinant Protein L, His Tag (RPL-PP2H2) is produced via site-specific conjugation of PE to Recombinant Protein L, His  Tag under optimal conditions with a proprietary technology. Recombinant Protein L is expressed from E.coli cells. It 1 * 200 Tests
Catalog # ACRBRPL-PP2H2-200
Supplier:  ACRO Biosystems
Frequently Bought Together