ContactUsLinkComponent
FITC-Labled Human Her2 / ErbB2, Fc Tag (HE2-HF256) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Acce ssion # P04626-1). 1 * 25 µG
Catalog # ACRBHE2-HF256-25UG
Supplier:  ACRO Biosystems
FITC-Labled Human Her2 / ErbB2, Fc Tag (HE2-HF256) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Acce ssion # P04626-1). 1 * 25 µG
Catalog # ACRBHE2-HF256-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together