ContactUsLinkComponent
Biotinylated Cynomolgus / Rhesus macaque PD-L1, His,Avitag (PDL-C82E8) is expressed from human 293 cells (HEK293). It contains AA Ph e 19 - Arg 238 (Accession # G7PSE7-1). 1 * 25 µG
Catalog # ACRBPDL-C82E8-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus / Rhesus macaque PD-L1, His,Avitag (PDL-C82E8) is expressed from human 293 cells (HEK293). It contains AA Ph e 19 - Arg 238 (Accession # G7PSE7-1). 1 * 25 µG
Catalog # ACRBPDL-C82E8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together