ContactUsLinkComponent
Human Claudin-18.2 Full Length Protein-VLP (CL2-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 261 (A ccession # P56856-2 ). 1 * 100 µG
Catalog # ACRBCL2-H52P7100UG
Supplier:  ACRO Biosystems
Human Claudin-18.2 Full Length Protein-VLP (CL2-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 261 (A ccession # P56856-2 ). 1 * 100 µG
Catalog # ACRBCL2-H52P7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together