ContactUsLinkComponent
Rhesus macaque Klotho beta, His Tag (KLB-R52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 53 - Leu 997 (Accessi on # EHH25814.1). 1 * 1 mg
Catalog # ACRBKLB-R52H8-1MG
Supplier:  ACROBIOSYSTEMS
Rhesus macaque Klotho beta, His Tag (KLB-R52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 53 - Leu 997 (Accessi on # EHH25814.1). 1 * 1 mg
Catalog # ACRBKLB-R52H8-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together