ContactUsLinkComponent
Rhesus macaque CA125 (13647-14291), His Tag (CA5-R52H6) is expressed from human 293 cells (HEK293). It contains AA Pro 13647 - Pro 1 4291 (Accession # XP_015296314.1). 1 * 1 mg
Catalog # ACRBCA5-R52H6-1MG
Supplier:  ACRO Biosystems
Rhesus macaque CA125 (13647-14291), His Tag (CA5-R52H6) is expressed from human 293 cells (HEK293). It contains AA Pro 13647 - Pro 1 4291 (Accession # XP_015296314.1). 1 * 1 mg
Catalog # ACRBCA5-R52H6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together