ContactUsLinkComponent
PE-Labeled Human uPAR, His Tag (UPR-HP2H7) is produced via site-specific conjugation of PE to Human uPAR, His Tag under optimal cond itions with a proprietary technology. Human uPAR, His Tag is expressed from human 293 cells (HEK293). It contains AA 1 * 200 Tests
Catalog # ACRBUPR-HP2H7-200
Supplier:  ACRO Biosystems
PE-Labeled Human uPAR, His Tag (UPR-HP2H7) is produced via site-specific conjugation of PE to Human uPAR, His Tag under optimal cond itions with a proprietary technology. Human uPAR, His Tag is expressed from human 293 cells (HEK293). It contains AA 1 * 200 Tests
Catalog # ACRBUPR-HP2H7-200
Supplier:  ACRO Biosystems
Frequently Bought Together